Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000929.1 | NP_001000929.1 | 1 | 279 |
Gene Symbol: Olr557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000669.1 | NP_001000669.1 | 1 | 378 |