Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000932.1 | NP_001000932.1 | 517 |
Gene Symbol: LOC100909831 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749506.1 | XP_003749554.1 | 1 | 517 |