Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004707.3 | NP_004698.3 | 2, 3 | 593 | ||
NR_033362.1 | 3, 4 | 710 | ||||
NR_033363.1 | 3, 4 | 709 | ||||
NR_073603.1 | 4, 5 | 826 | ||||
GenBank mRNA | AB017507.1 | - | 2, 3 | 313 | ||
AK098251.1 | - | 2, 3 | 1763 | |||
AK313320.1 | - | 2, 3 | 588 | |||
BC012266.2 | - | 2, 3 | 544 | 3864058 | ||
CB995198.1 | - | 3, 4 | 421 | |||
CN400030.1 | - | 3, 4 | 344 | |||
CR978239.1 | - | 4, 5 | 519 | |||
EU554057.1 | - | 2, 3 | 435 |