Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000945.1 | NP_001000945.1 | 1 | 393 |
Gene Symbol: Olr1545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001105.1 | NP_001001105.1 | 1 | 387 |
Gene Symbol: Olr1546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001106.1 | NP_001001106.1 | 1 | 387 |
Gene Symbol: Olr1547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001107.1 | NP_001001107.1 | 1 | 387 |