Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001681.3 | NP_001672.1 | 13 | 2200 | ||
NM_170665.3 | NP_733765.1 | 13 | 2200 | |||
XM_005253888.2 | XP_005253945.1 | 13 | 2195 | |||
XM_011538402.2 | XP_011536704.1 | 13 | 2195 | |||
GenBank mRNA | AK293877.1 | - | 9 | 1308 | ||
BC035588.1 | - | 13 | 1747 | 5503508 | ||
KU177929.1 | - | 13 | 1637 | |||
M23114.1 | - | 13 | 1800 | |||
M23115.1 | - | 13 | 1640 |