Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000963.1 | NP_001000963.1 | 1 | 329 |
Gene Symbol: LOC100911939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749657.1 | XP_003749705.1 | 1 | 329 |
Gene Symbol: LOC103692039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762673.1 | XP_008760895.1 | 1 | 329 |
Gene Symbol: Olr1238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001013.1 | NP_001001013.1 | 1 | 329 |