Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320523.1 | NP_001307452.1 | 2 | 226 | ||
NM_004710.4 | NP_004701.1 | 2 | 226 | |||
XM_005257792.3 | XP_005257849.1 | 2 | 220 | |||
GenBank mRNA | AJ002308.1 | - | 2 | 183 | ||
AL833010.1 | - | 1 | 1188 | |||
AY358916.1 | - | 2 | 204 | |||
BC000407.2 | - | 2 | 163 | 2823026 | ||
BC029755.1 | - | 2 | 159 | 4341106 | ||
BC105992.1 | - | 2 | 160 | 4746277 | ||
CF122406.1 | - | 2 | 157 | |||
HY054610.1 | - | 2 | 226 |