Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001002.1 | NP_001001002.1 | 1 | 43 |
Gene Symbol: Olr1416 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000009.1 | NP_001000009.1 | 1 | 43 |
Gene Symbol: Olr1418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000008.1 | NP_001000008.1 | 1 | 43 |
Gene Symbol: Olr1421 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000780.1 | NP_001000780.1 | 1 | 61 |