Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEURL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004210.4 | NP_004201.3 | 4, 5 | 1749 | ||
XM_005270269.3 | XP_005270326.1 | 4, 5 | 1418 | |||
XM_005270270.3 | XP_005270327.1 | 4, 5 | 1395 | |||
XM_011540331.2 | XP_011538633.1 | 5, 6 | 1951 | |||
XM_011540332.2 | XP_011538634.1 | 5, 6 | 1598 | |||
XM_011540333.2 | XP_011538635.1 | 4, 5 | 1484 | |||
XM_017016909.1 | XP_016872398.1 | 4, 5 | 1378 | |||
GenBank mRNA | AF029729.1 | - | 4, 5 | 1400 | ||
AK299620.1 | - | 1, 2 | 268 | |||
BC026336.1 | - | 4, 5 | 1436 | 4812302 | ||
U87864.1 | - | 4, 5 | 1749 |