Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202504.1 | NP_001189433.1 | 8 | 1722 | ||
NM_001318511.1 | NP_001305440.1 | 8 | 2095 | |||
NM_004715.4 | NP_004706.3 | 8 | 2095 | |||
NM_048368.3 | NP_430255.2 | 8 | 2095 | |||
XM_011526261.1 | XP_011524563.1 | 8 | 2073 | |||
XM_017026078.1 | XP_016881567.1 | 6 | 1575 | |||
GenBank mRNA | AF081287.1 | - | 7 | 1643 | ||
AF154115.1 | - | 8 | 2031 | |||
BC015010.1 | - | 2 | 1042 | |||
BC032515.1 | - | 8 | 1749 | |||
BC047999.1 | - | 8 | 2095 | |||
BC052576.1 | - | 8 | 1687 | |||
BC063447.1 | - | 8 | 2050 | 5500032 |