Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC28A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004212.3 | NP_004203.2 | 16 | 1725 | ||
XM_011522198.2 | XP_011520500.1 | 16 | 1745 | |||
XM_011522202.2 | XP_011520504.1 | 8 | 896 | |||
GenBank mRNA | AF036109.1 | - | 15 | 1674 | ||
AK025121.1 | - | 1 | 533 | |||
AK222709.1 | - | 16 | 1725 | |||
AK291974.1 | - | 16 | 1696 | |||
BC093737.1 | - | 15 | 1688 | 7939582 | ||
BC112047.1 | - | 15 | 1700 | 8327515 | ||
U84392.1 | - | 16 | 1719 |