Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001064.1 | NP_001001064.1 | 1 | 249 |
Gene Symbol: Olr698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000513.1 | NP_001000513.1 | 1 | 249 |