Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004214.4 | NP_004205.2 | 6 | 791 | ||
NM_198897.1 | NP_942600.1 | 6 | 812 | |||
GenBank mRNA | AF010187.2 | - | 6 | 688 | ||
AF171944.1 | - | 6 | 670 | |||
AF171945.1 | - | 6 | 670 | |||
AF171946.1 | - | 6 | 670 | |||
AF250391.1 | - | 6 | 691 | |||
AK223438.1 | - | 6 | 713 | |||
AK289562.1 | - | 6 | 713 | |||
BC014388.2 | - | 6 | 731 | 3504921 | ||
BC017448.1 | - | 6 | 687 | 3509570 | ||
CR456749.1 | - | 6 | 670 |