Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004716.3 | NP_004707.2 | 12 | 2108 | ||
XM_006718940.3 | XP_006719003.1 | 5 | 867 | |||
XM_017018546.1 | XP_016874035.1 | 7 | 636 | |||
GenBank mRNA | AK027156.1 | - | 1 | 446 | ||
AK097207.1 | - | 4 | 1283 | |||
AK222907.1 | - | 12 | 1648 | |||
AK312429.1 | - | 12 | 1683 | |||
BC006357.2 | - | 12 | 1626 | 4125594 | ||
BC010696.1 | - | 12 | 1678 | 3868257 | ||
BT006870.1 | - | 10 | 1477 | |||
U33849.1 | - | 12 | 1628 | |||
U40623.1 | - | 10 | 1498 |