Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001096.1 | NP_001001096.1 | 248 |
Gene Symbol: Olr1399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001095.1 | NP_001001095.1 | 1 | 248 |
Gene Symbol: Olr1401 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000003.1 | NP_001000003.1 | 1 | 248 |