Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321708.1 | NP_001308637.1 | 21 | 2190 | ||
NM_001321709.1 | NP_001308638.1 | 22 | 1968 | |||
NM_001321710.1 | NP_001308639.1 | 21 | 2156 | |||
NM_004717.3 | NP_004708.1 | 21 | 2190 | |||
XM_011516682.2 | XP_011514984.2 | 22 | 2250 | |||
XM_011516684.2 | XP_011514986.2 | 15 | 9434 | |||
XM_017012780.1 | XP_016868269.1 | 22 | 2251 | |||
XM_017012781.1 | XP_016868270.1 | 22 | 2235 | |||
XM_017012782.1 | XP_016868271.1 | 21 | 2136 | |||
XM_017012783.1 | XP_016868272.1 | 22 | 2250 | |||
XM_017012784.1 | XP_016868273.1 | 20 | 2061 | |||
XM_017012785.1 | XP_016868274.1 | 20 | 2438 | |||
XM_017012786.1 | XP_016868275.1 | 22 | 1938 | |||
XM_017012787.1 | XP_016868276.1 | 21 | 1803 | |||
XM_017012788.1 | XP_016868277.1 | 22 | 1921 | |||
XM_017012789.1 | XP_016868278.1 | 8 | 647 | |||
GenBank mRNA | AB209167.1 | - | 21 | 1828 |