Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001100.1 | NP_001001100.1 | 1 | 427 | ||
XM_006248195.2 | XP_006248257.1 | 2 | 526 |
Gene Symbol: Olr1530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001101.1 | NP_001001101.1 | 1 | 427 | ||
GenBank mRNA | AF091575.1 | - | 1 | 241 |
Gene Symbol: Olr1531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001102.1 | NP_001001102.1 | 1 | 427 |
Gene Symbol: Olr1532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001103.1 | NP_001001103.1 | 1 | 427 |