Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001102.1 | NP_001001102.1 | 1 | 560 |
Gene Symbol: Olr1532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001103.1 | NP_001001103.1 | 1 | 560 |