Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001112.1 | NP_001001112.1 | 1 | 340 |
Gene Symbol: LOC100362856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728918.3 | XP_002728964.3 | 1 | 340 |
Gene Symbol: Olr1696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206850.1 | NP_996732.1 | 1 | 340 | ||
GenBank mRNA | AF034896.1 | - | 562 |
Gene Symbol: Olr1697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001111.1 | NP_001001111.1 | 1 | 340 |