Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001113.1 | NP_001001113.1 | 1 | 126 |
Gene Symbol: LOC103689944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255858.2 | XP_006255920.1 | 1 | 126 |
Gene Symbol: Olr1687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000508.1 | NP_001000508.1 | 1 | 126 |
Gene Symbol: Olr1696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206850.1 | NP_996732.1 | 1 | 126 | ||
GenBank mRNA | AF034896.1 | - | 348 |
Gene Symbol: Olr1697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001111.1 | NP_001001111.1 | 1 | 126 |