Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001113.1 | NP_001001113.1 | 1 | 641 |
Gene Symbol: LOC100910739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751909.2 | XP_003751957.1 | 1 | 452 |
Gene Symbol: LOC103689944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255858.2 | XP_006255920.1 | 1 | 479 |
Gene Symbol: Olr1701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001114.1 | NP_001001114.1 | 1 | 641 | ||
XM_006255861.2 | XP_006255923.1 | 2 | 832 | |||
XM_006255863.2 | XP_006255925.1 | 2 | 801 | |||
XM_006255865.2 | XP_006255927.1 | 1 | 647 |