Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7A2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319036.1 | NP_001305965.1 | 4 | 1703 | ||
NM_001319037.1 | NP_001305966.1 | 3 | 944 | |||
NM_001319040.1 | NP_001305969.1 | 3 | 1004 | |||
NM_004718.3 | NP_004709.2 | 3 | 1008 | |||
NR_134947.1 | 3 | 1160 | ||||
NR_134948.1 | 3 | 1787 | ||||
GenBank mRNA | AB007618.1 | - | 3 | 980 | ||
AK092911.1 | - | 4 | 1755 | |||
AK130281.1 | - | 3 | 1797 | |||
BC005251.1 | - | 3 | 971 | 3930624 | ||
BC007095.1 | - | 3 | 991 | 4251503 |