Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7A2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319036.1 | NP_001305965.1 | 3 | 972 | ||
NM_001319037.1 | NP_001305966.1 | 2 | 213 | |||
NM_004718.3 | NP_004709.2 | 2 | 277 | |||
NR_134947.1 | 2 | 429 | ||||
NR_134948.1 | 2 | 1056 | ||||
GenBank mRNA | AB007618.1 | - | 2 | 235 | ||
AF127788.1 | - | 2 | 195 | |||
AK092911.1 | - | 3 | 1025 | |||
AK130281.1 | - | 2 | 1056 | |||
BC005251.1 | - | 2 | 240 | 3930624 | ||
BC007095.1 | - | 2 | 250 | 4251503 | ||
BF130603.1 | - | 2 | 277 | |||
BI598394.1 | - | 2 | 382 | |||
BT007371.1 | - | 2 | 195 | |||
CF593726.1 | - | 2 | 241 | |||
CR542208.1 | - | 2 | 195 |