Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001331.2 | NP_001001331.1 | 23 | 4055 | ||
NM_001683.3 | NP_001674.2 | 20 | 3920 | |||
XM_005265179.4 | XP_005265236.1 | 25 | 4239 | |||
XM_006713175.3 | XP_006713238.1 | 25 | 4123 | |||
XM_011533752.2 | XP_011532054.1 | 23 | 4028 | |||
XM_017006481.1 | XP_016861970.1 | 26 | 4366 | |||
XM_017006482.1 | XP_016861971.1 | 23 | 4057 | |||
XM_017006483.1 | XP_016861972.1 | 21 | 4067 | |||
XM_017006484.1 | XP_016861973.1 | 23 | 4030 | |||
XM_017006485.1 | XP_016861974.1 | 24 | 4334 | |||
XM_017006486.1 | XP_016861975.1 | 25 | 4389 | |||
XM_017006487.1 | XP_016861976.1 | 22 | 3987 | |||
XM_017006488.1 | XP_016861977.1 | 22 | 4241 | |||
XM_017006489.1 | XP_016861978.1 | 23 | 4163 | |||
XM_017006490.1 | XP_016861979.1 | 21 | 4081 | |||
XM_017006491.1 | XP_016861980.1 | 22 | 4253 | |||
XM_017006492.1 | XP_016861981.1 | 24 | 4248 | |||
GenBank mRNA | AB210008.1 | - | 20 | 3993 | ||
L00620.1 | - | 19 | 3499 | |||
L20977.1 | - | 22 | 4057 | |||
M97260.1 | - | 20 | 3901 | |||
S95758.1 | - | 19 | 3184 | |||
X63575.1 | - | 22 | 3686 |