Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212458.1 | NP_997623.1 | 3 | 482 | ||
XM_006255972.1 | XP_006256034.1 | 3 | 335 | |||
XM_006255973.1 | XP_006256035.1 | 2 | 236 | |||
XM_006255974.1 | XP_006256036.1 | 3 | 424 | |||
XM_006255975.1 | XP_006256037.1 | 3 | 725 | |||
GenBank mRNA | BC085852.1 | - | 3 | 428 | 7189891 |