Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003970.3 | NP_003961.3 | 26 | 3377 | ||
XM_006716237.1 | XP_006716300.1 | 26 | 3342 | |||
GenBank mRNA | AK297890.1 | - | 13 | 1695 | ||
BC052969.1 | - | 26 | 3330 | 6205359 | ||
BP260135.1 | - | 1 | 50 | |||
BP260324.1 | - | 1 | 54 | |||
BX501741.1 | - | 6 | 566 | |||
DA900083.1 | - | 5 | 367 | |||
X69089.1 | - | 26 | 3287 |