Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749452.3 | XP_003749500.2 | 6 | 1052 | ||
XM_006234012.2 | XP_006234074.1 | 5 | 1214 | |||
XM_008761787.1 | XP_008760009.1 | 4 | 1028 | |||
GenBank mRNA | AJ646871.1 | - | 4 | 783 |