Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002834.1 | NP_001002834.1 | 1 | 1256 |
Gene Symbol: LOC683408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749847.3 | XP_003749895.3 | 1 | 230 |
Gene Symbol: Senp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002833.1 | NP_001002833.1 | 1256 |