Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001331.2 | NP_001001331.1 | 4 | 1001 | ||
NM_001683.3 | NP_001674.2 | 4 | 1001 | |||
XM_005265179.4 | XP_005265236.1 | 6 | 1185 | |||
XM_006713175.3 | XP_006713238.1 | 6 | 1069 | |||
XM_011533752.2 | XP_011532054.1 | 4 | 974 | |||
XM_017006481.1 | XP_016861970.1 | 7 | 1312 | |||
XM_017006482.1 | XP_016861971.1 | 4 | 1003 | |||
XM_017006483.1 | XP_016861972.1 | 4 | 1106 | |||
XM_017006484.1 | XP_016861973.1 | 6 | 1069 | |||
XM_017006485.1 | XP_016861974.1 | 4 | 1108 | |||
XM_017006486.1 | XP_016861975.1 | 4 | 1108 | |||
XM_017006487.1 | XP_016861976.1 | 6 | 1068 | |||
XM_017006488.1 | XP_016861977.1 | 4 | 1108 | |||
XM_017006489.1 | XP_016861978.1 | 4 | 975 | |||
XM_017006490.1 | XP_016861979.1 | 4 | 990 | |||
XM_017006491.1 | XP_016861980.1 | 4 | 1107 | |||
XM_017006492.1 | XP_016861981.1 | 4 | 1107 | |||
GenBank mRNA | AB210008.1 | - | 4 | 1074 | ||
AK308898.1 | - | 4 | 1001 | |||
L00620.1 | - | 3 | 580 | |||
L20977.1 | - | 6 | 1138 | |||
M97260.1 | - | 4 | 982 | |||
S95738.1 | - | 3 | 632 | |||
S95743.1 | - | 2 | 378 | |||
S95758.1 | - | 2 | 223 | |||
X63575.1 | - | 3 | 632 |