Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004094.1 | NP_001004094.1 | 9, 10 | 683 | ||
GenBank mRNA | M58593.1 | - | 9, 10 | 683 |
Gene Symbol: Psma3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017280.2 | NP_058976.1 | 9, 10 | 658 | ||
GenBank mRNA | BC081817.1 | - | 9, 10 | 658 | 7110009 | |
D90258.1 | - | 9, 10 | 683 | |||
FQ211111.1 | - | 9, 10 | 687 | |||
FQ219582.1 | - | 9, 10 | 778 | |||
FQ224319.1 | - | 685 | ||||
FQ233787.1 | - | 9, 10 | 720 | |||
X55985.1 | - | 9, 10 | 694 |