Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nanog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100781.1 | NP_001094251.1 | 4 | 784 | ||
XM_006237310.2 | XP_006237372.1 | 4 | 578 | |||
GenBank mRNA | AB162852.1 | - | 3 | 385 | ||
AB275459.1 | - | 4 | 578 |