Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-CE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008833.1 | NP_001008833.1 | 5 | 922 | ||
XM_006255847.2 | XP_006255909.1 | 5 | 927 | |||
GenBank mRNA | BC083874.1 | - | 4 | 1106 | ||
BC098952.1 | - | 5 | 925 | 7366301 |
Gene Symbol: RT1-A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008830.1 | NP_001008830.1 | 6 | 913 |
Gene Symbol: RT1-EC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X90374.1 | - | 4 | 850 |