Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136151.1 | NP_001129623.1 | 1 | 892 | ||
GenBank mRNA | D89995.1 | - | 1 | 738 | ||
D89996.1 | - | 1 | 892 |