Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003959.1 | NP_001003959.1 | 3 | 475 | ||
XM_006235340.2 | XP_006235402.1 | 2 | 345 | |||
XM_006235342.2 | XP_006235404.1 | 2 | 468 | |||
XM_006235347.2 | XP_006235409.1 | 2 | 345 | |||
XM_008762364.1 | XP_008760586.1 | 1 | 1747 | |||
XM_008762365.1 | XP_008760587.1 | 1 | 1747 | |||
XM_008762366.1 | XP_008760588.1 | 1 | 1747 | |||
XM_008762367.1 | XP_008760589.1 | 1 | 1747 | |||
XM_008762368.1 | XP_008760590.1 | 2 | 125 | |||
XM_008762369.1 | XP_008760591.1 | 1 | 1747 | |||
XM_008762370.1 | XP_008760592.1 | 1 | 1747 | |||
XM_008762371.1 | XP_008760593.1 | 1 | 1747 | |||
XM_008762372.1 | XP_008760594.1 | 1 | 1747 |