Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dear | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004448.1 | NP_001004448.1 | 1 | 2985 | ||
GenBank mRNA | AY664492.1 | - | 1 | 2985 |