Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008398.2 | NP_001008399.1 | 2 | 687 | ||
GenBank mRNA | AJ633684.1 | - | 2 | 848 | ||
BC103635.1 | - | 2 | 687 | 7100941 | ||
CO556990.1 | - | 1 | 68 | |||
DQ125337.1 | - | 2 | 686 | |||
DQ125338.1 | - | 2 | 686 |