Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008398.2 | NP_001008399.1 | 2 | 508 | ||
GenBank mRNA | AJ633684.1 | - | 2 | 669 | ||
BC103635.1 | - | 2 | 508 | 7100941 | ||
DQ125337.1 | - | 2 | 507 | |||
DQ125338.1 | - | 2 | 507 |