Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008527.1 | NP_001008527.1 | 27 | 3177 | ||
XM_006245004.2 | XP_006245066.1 | 27 | 3205 | |||
XM_008767121.1 | XP_008765343.1 | 27 | 3206 | |||
XM_008767122.1 | XP_008765344.1 | 23 | 3874 | |||
XM_008767123.1 | XP_008765345.1 | 21 | 2696 | |||
XM_008767124.1 | XP_008765346.1 | 22 | 2872 | |||
GenBank mRNA | AY665586.1 | - | 27 | 3177 |