Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009492.1 | NP_001009492.1 | 4 | 830 | ||
XM_008769248.1 | XP_008767470.1 | 4 | 831 | |||
GenBank mRNA | AY196698.1 | - | 4 | 830 |