Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly49si3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009919.1 | NP_001009919.1 | 3 | 237 | ||
XM_006237116.2 | XP_006237178.1 | 331 | ||||
XM_008763410.1 | XP_008761632.1 | 4 | 518 | |||
XM_008763411.1 | XP_008761633.1 | 4 | 1155 | |||
XM_008763412.1 | XP_008761634.1 | 4 | 312 | |||
XM_008763413.1 | XP_008761635.1 | 4 | 518 | |||
XM_008763414.1 | XP_008761636.1 | 4 | 518 | |||
XM_008763415.1 | XP_008761637.1 | 4 | 518 | |||
GenBank mRNA | AY653730.1 | - | 3 | 237 |
Gene Symbol: LOC100910264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763346.1 | XP_008761568.1 | 3 | 2816 |
Gene Symbol: Ly49si2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009498.1 | NP_001009498.1 | 237 | |||
XM_008763406.1 | XP_008761628.1 | 250 | ||||
XM_008763407.1 | XP_008761629.1 | 250 | ||||
XM_008763408.1 | XP_008761630.1 | 250 | ||||
XM_008763409.1 | XP_008761631.1 | 250 | ||||
GenBank mRNA | AY659933.1 | - | 237 |