Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009508.1 | NP_001009508.1 | 1 | 123 | ||
GenBank mRNA | AY510284.1 | - | 1 | 123 |
Gene Symbol: Vom1r73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008922.1 | NP_001008922.1 | 1 | 180 | ||
GenBank mRNA | AY510289.1 | - | 1 | 180 |