Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009524.1 | NP_001009524.1 | 6 | 787 | ||
XM_006242737.2 | XP_006242799.1 | 6 | 665 | |||
GenBank mRNA | AF299297.1 | - | 6 | 787 | ||
AF299298.1 | - | 6 | 641 | |||
AF414431.1 | - | 6 | 641 | |||
AY040870.1 | - | 6 | 787 |