Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009528.1 | NP_001009528.1 | 1 | 198 | ||
XM_008762962.1 | XP_008761184.1 | 2 | 271 | |||
GenBank mRNA | AY510312.1 | - | 1 | 198 |
Gene Symbol: Vom1r86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009527.1 | NP_001009527.1 | 1 | 198 | ||
GenBank mRNA | AY510308.1 | - | 1 | 198 |