Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008902.1 | NP_001008902.1 | 1 | 615 | ||
XM_008758769.1 | XP_008756991.1 | 3 | 1637 | |||
XM_008758770.1 | XP_008756992.1 | 4 | 1159 | |||
GenBank mRNA | AY510307.1 | - | 1 | 615 |