Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008909.1 | NP_001008909.1 | 1 | 559 | ||
GenBank mRNA | AY510299.1 | - | 1 | 559 |
Gene Symbol: Vom1r107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008908.1 | NP_001008908.1 | 1 | 559 | ||
GenBank mRNA | AY510300.1 | - | 1 | 559 |