Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008928.1 | NP_001008928.1 | 1 | 88 | ||
GenBank mRNA | AY510298.1 | - | 1 | 88 |