Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008951.1 | NP_001008951.1 | 1 | 770 | ||
GenBank mRNA | AY510278.1 | - | 1 | 770 |