Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008955.1 | NP_001008955.1 | 1 | 379 | ||
GenBank mRNA | AY510291.1 | - | 1 | 379 |
Gene Symbol: LOC103690997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758821.1 | XP_008757043.1 | 406 |