Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171162.1 | NP_001164633.1 | 2 | 1366 | ||
NM_001171163.1 | NP_001164634.1 | 2 | 805 | |||
NM_005096.3 | NP_005087.1 | 2 | 805 | |||
NM_201599.2 | NP_963893.1 | 2 | 796 | |||
XM_005262309.3 | XP_005262366.1 | 2 | 1239 | |||
XM_005262310.2 | XP_005262367.1 | 2 | 752 | |||
XM_011531062.2 | XP_011529364.1 | 2 | 705 | |||
GenBank mRNA | AB002383.1 | - | 2 | 704 | ||
AK290762.1 | - | 2 | 805 | |||
BC013009.2 | - | 2 | 729 | |||
BC069057.1 | - | 2 | 709 | 5499547 | ||
BT007095.1 | - | 1 | 617 | |||
X95808.1 | - | 3 | 1342 |