Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008962.1 | NP_001008962.1 | 1 | 472 | ||
GenBank mRNA | AY510364.1 | - | 1 | 472 |
Gene Symbol: Vom1r15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008960.1 | NP_001008960.1 | 1 | 472 | ||
GenBank mRNA | AY510361.1 | - | 1 | 472 |